2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide

Lieferant: Apollo Scientific

PC410345-1G
APOSPC410345-1GEA 386 EUR
APOSPC410345-1G
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Formel: C₈H₁₁F₃N₄O
Molecular Weight: 236,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419690
CAS-Nummer: 1005592-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR