1-(2,3,5,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine
Lieferant: Apollo Scientific
PC410347-1G
APOSPC410347-1GEA
516
EUR
APOSPC410347-1G
1-(2,3,5,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine
1-(2,3,5,6-Tetrafluorobenzyl)-1H-pyrazol-4-amine
Formel:
C₁₀H₇F₄N₃ Molecular Weight: 245,176 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04967238 CAS-Nummer: 1005630-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...