2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanehydrazide

Lieferant: Apollo Scientific

PC410348-1G
APOSPC410348-1GEA 0 EUR
APOSPC410348-1G
2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanehydrazide
2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanehydrazide
Formel: C₈H₁₂F₂N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD03419705
CAS-Nummer: 1005631-76-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR