2-[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific

PC410349-1G
APOSPC410349-1GEA 647 EUR
APOSPC410349-1G
2-[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid
2-[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid
Formel: C₈H₉BrF₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03419701
CAS-Nummer: 1005631-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR