2-[4-Bromo-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide

Lieferant: Apollo Scientific

PC410350-1G
APOSPC410350-1GEA 516 EUR
APOSPC410350-1G
2-[4-Bromo-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
2-[4-Bromo-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
Formel: C₉H₁₁BrF₂N₄O
Molecular Weight: 309.111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04969901
CAS-Nummer: 1005631-88-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR