3-Amino-4-(1,5-dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide

Lieferant: Apollo Scientific

PC410352-1G
APOSPC410352-1GEA 516 EUR
APOSPC410352-1G
3-Amino-4-(1,5-dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
3-Amino-4-(1,5-dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
Formel: C₁₄H₁₂F₃N₅OS
Molecular Weight: 355.338 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422767
CAS-Nummer: 1005632-64-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR