2-({4-Ethyl-5-[1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]-4H-1,2,4-triazol-3-yl}sulphanyl)acetohydrazide

Lieferant: Apollo Scientific

PC410353-1G
APOSPC410353-1GEA 516 EUR
APOSPC410353-1G
2-({4-Ethyl-5-[1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]-4H-1,2,4-triazol-3-yl}sulphanyl)acetohydrazide
2-({4-Ethyl-5-[1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]-4H-1,2,4-triazol-3-yl}sulphanyl)acetohydrazide
Formel: C₁₁H₁₄F₃N₇OS
Molecular Weight: 349.335 g/mol
MDL: MFCD03423243
CAS-Nummer: 1005632-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR