2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Lieferant: Apollo Scientific
PC410355-1G
APOSPC410355-1GEA
386
EUR
APOSPC410355-1G
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Formel:
C₁₀H₁₃F₃N₄O Molecular Weight: 262.232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419748 CAS-Nummer: 1005650-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...