2-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide

Lieferant: Apollo Scientific

PC410356-1G
APOSPC410356-1GEA 386 EUR
APOSPC410356-1G
2-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
2-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Formel: C₈H₁₀ClF₃N₄O
Molecular Weight: 270,639 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419691
CAS-Nummer: 1005651-08-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR