2-[4-Chloro-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific

PC410358-1G
APOSPC410358-1GEA 828 EUR
APOSPC410358-1G
2-[4-Chloro-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]propanoic acid
2-[4-Chloro-3,5-bis(difluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel: C₈H₇ClF₄N₂O₂
Molecular Weight: 274,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419716
CAS-Nummer: 1005678-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR