2-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific

PC410359-1G
APOSPC410359-1GEA 516 EUR
APOSPC410359-1G
2-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
2-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel: C₈H₈ClF₃N₂O₂
Molecular Weight: 256,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419688
CAS-Nummer: 1005679-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR