3-Amino-6-(difluoromethyl)-4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide

Lieferant: Apollo Scientific

PC410360-1G
APOSPC410360-1GEA 516 EUR
APOSPC410360-1G
3-Amino-6-(difluoromethyl)-4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
3-Amino-6-(difluoromethyl)-4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
Formel: C₁₅H₁₅F₂N₅OS
Molecular Weight: 351,374 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422786
CAS-Nummer: 1005679-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR