3-Amino-6-(difluoromethyl)-4-(1,3-dimethyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide

Lieferant: Apollo Scientific

PC410361-1G
APOSPC410361-1GEA 516 EUR
APOSPC410361-1G
3-Amino-6-(difluoromethyl)-4-(1,3-dimethyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
3-Amino-6-(difluoromethyl)-4-(1,3-dimethyl-1H-pyrazol-4-yl)thieno[2,3-b]pyridine-2-carboxamide
Formel: C₁₄H₁₃F₂N₅OS
Molecular Weight: 337,348 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422784
CAS-Nummer: 1005694-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR