3-Amino-4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
Lieferant: Apollo Scientific
PC410363-1G
APOSPC410363-1GEA
516
EUR
APOSPC410363-1G
3-Amino-4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
3-Amino-4-(1-ethyl-3-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide
Formel:
C₁₅H₁₄F₃N₅OS Molecular Weight: 369,365 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03422769 CAS-Nummer: 1005696-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...