3-[1'-Methyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propan-1-amine

Lieferant: Apollo Scientific

PC410366-1G
APOSPC410366-1GEA 516 EUR
APOSPC410366-1G
3-[1'-Methyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propan-1-amine
3-[1'-Methyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propan-1-amine
Formel: C₁₁H₁₄F₃N₅
Molecular Weight: 273,258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558972
CAS-Nummer: 1006319-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR