3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-2-methylpropanenitrile
Lieferant: Apollo Scientific
PC410367-1G
APOSPC410367-1GEA
386
EUR
APOSPC410367-1G
3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-2-methylpropanenitrile
3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-2-methylpropanenitrile
Formel:
C₁₁H₁₂F₃N₃ Molecular Weight: 243,228 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08558989 CAS-Nummer: 1006319-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...