[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid
Lieferant: Apollo Scientific
PC410368-1G
APOSPC410368-1GEA
0
EUR
APOSPC410368-1G
[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid
[4-Bromo-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid
Formel:
C₆H₄BrF₃N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04969233 CAS-Nummer: 1006319-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...