3-(3-Trifluoromethylpyrazol-1-yl)propionic acid
Lieferant: Apollo Scientific
PC410369-1G
APOSPC410369-1GEA
232
EUR
APOSPC410369-1G
3-(3-Trifluoromethylpyrazol-1-yl)propionic acid
3-(3-Trifluoromethylpyrazol-1-yl)propionic acid
Formel:
C₇H₇F₃N₂O₂ Molecular Weight: 208,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04969242 CAS-Nummer: 1006319-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...