3-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]butanenitrile

Lieferant: Apollo Scientific

PC410371-1G
APOSPC410371-1GEA 516 EUR
APOSPC410371-1G
3-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]butanenitrile
3-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]butanenitrile
Formel: C₈H₈F₃N₃
Molecular Weight: 203.164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558149
CAS-Nummer: 1006319-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR