3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile

Lieferant: Apollo Scientific

PC410372-1G
APOSPC410372-1GEA 386 EUR
APOSPC410372-1G
3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Formel: C₁₀H₁₀F₃N₃
Molecular Weight: 229.202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558159
CAS-Nummer: 1006319-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR