3-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]pentan-1-amine

Lieferant: Apollo Scientific

PC410373-1G
APOSPC410373-1GEA 516 EUR
APOSPC410373-1G
3-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]pentan-1-amine
3-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]pentan-1-amine
Formel: C₉H₁₄F₃N₃
Molecular Weight: 221.223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06805065
CAS-Nummer: 1006320-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR