3-[1'-Ethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propanenitrile
Lieferant: Apollo Scientific
PC410374-1G
APOSPC410374-1GEA
386
EUR
APOSPC410374-1G
3-[1'-Ethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propanenitrile
3-[1'-Ethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propanenitrile
Formel:
C₁₂H₁₂F₃N₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08558461 CAS-Nummer: 1006320-21-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...