2-[4-Bromo-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Lieferant: Apollo Scientific
PC410375-1G
APOSPC410375-1GEA
647
EUR
APOSPC410375-1G
2-[4-Bromo-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
2-[4-Bromo-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel:
C₁₀H₁₀BrF₃N₂O₂ Molecular Weight: 327,098 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419747 CAS-Nummer: 1006320-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...