1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-4,4,4-trifluorobutane-1,3-dione

Lieferant: Apollo Scientific

PC410376-1G
APOSPC410376-1GEA 516 EUR
APOSPC410376-1G
1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-4,4,4-trifluorobutane-1,3-dione
1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-4,4,4-trifluorobutane-1,3-dione
Formel: C₁₀H₁₁F₃N₂O₂
Molecular Weight: 248,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419774
CAS-Nummer: 1006322-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR