(1Z)-3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-N'-hydroxypropanimidamide
Lieferant: Apollo Scientific
PC410377-1G
APOSPC410377-1GEA
647
EUR
APOSPC410377-1G
(1Z)-3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-N'-hydroxypropanimidamide
(1Z)-3-[3-Cyclopropyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-N'-hydroxypropanimidamide
Formel:
C₁₀H₁₃F₃N₄O Molecular Weight: 262,232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15976232 CAS-Nummer: 1006322-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...