3-[1',3'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propan-1-amine
Lieferant: Apollo Scientific
PC410378-1G
APOSPC410378-1GEA
516
EUR
APOSPC410378-1G
3-[1',3'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propan-1-amine
3-[1',3'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propan-1-amine
Formel:
C₁₂H₁₆F₃N₅ Molecular Weight: 287,284 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08556263 CAS-Nummer: 1006323-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...