2-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide

Lieferant: Apollo Scientific

PC410379-1G
APOSPC410379-1GEA 386 EUR
APOSPC410379-1G
2-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
2-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Formel: C₇H₉F₃N₄O
Molecular Weight: 222,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04969239
CAS-Nummer: 1006327-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR