2-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Lieferant: Apollo Scientific
PC410380-1G
APOSPC410380-1GEA
516
EUR
APOSPC410380-1G
2-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
2-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel:
C₇H₇F₃N₂O₂ Molecular Weight: 208,138 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04969236 CAS-Nummer: 1006328-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...