1-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propan-2-amine

Lieferant: Apollo Scientific

PC410381-1G
APOSPC410381-1GEA 647 EUR
APOSPC410381-1G
1-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propan-2-amine
1-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]propan-2-amine
Formel: C₇H₁₀F₃N₃
Molecular Weight: 193,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970168
CAS-Nummer: 1006328-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR