4-(1,3-Dimethyl-1H-pyrazol-4-yl)-2-sulphanyl-6-(trifluoromethyl)pyridine-3-carbonitrile

Lieferant: Apollo Scientific

PC410382-1G
APOSPC410382-1GEA 647 EUR
APOSPC410382-1G
4-(1,3-Dimethyl-1H-pyrazol-4-yl)-2-sulphanyl-6-(trifluoromethyl)pyridine-3-carbonitrile
4-(1,3-Dimethyl-1H-pyrazol-4-yl)-2-sulphanyl-6-(trifluoromethyl)pyridine-3-carbonitrile
Formel: C₁₂H₉F₃N₄S
Molecular Weight: 298.287 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422724
CAS-Nummer: 1006333-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR