2-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Lieferant: Apollo Scientific
PC410383-1G
APOSPC410383-1GEA
647
EUR
APOSPC410383-1G
2-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
2-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Formel:
C₈H₈ClF₃N₂O₂ Molecular Weight: 256,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970064 CAS-Nummer: 1006333-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...