3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanenitrile

Lieferant: Apollo Scientific

PC410385-1G
APOSPC410385-1GEA 516 EUR
APOSPC410385-1G
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanenitrile
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanenitrile
Formel: C₁₁H₁₂F₃N₃
Molecular Weight: 243,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558137
CAS-Nummer: 1006333-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR