3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile

Lieferant: Apollo Scientific

PC410386-1G
APOSPC410386-1GEA 516 EUR
APOSPC410386-1G
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Formel: C₁₀H₁₀F₃N₃
Molecular Weight: 229,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558158
CAS-Nummer: 1006334-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR