{2-Methyl-3-[3-(trifluoromethyl)-1H-pyrazol-1-yl]propyl}amine
Lieferant: Apollo Scientific
PC410387-1G
APOSPC410387-1GEA
516
EUR
APOSPC410387-1G
{2-Methyl-3-[3-(trifluoromethyl)-1H-pyrazol-1-yl]propyl}amine
{2-Methyl-3-[3-(trifluoromethyl)-1H-pyrazol-1-yl]propyl}amine
Formel:
C₈H₁₂F₃N₃ Molecular Weight: 207,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06805050 CAS-Nummer: 1006334-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...