1-[4-(1,3-Dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid
Lieferant: Apollo Scientific
PC410389-1G
APOSPC410389-1GEA
516
EUR
APOSPC410389-1G
1-[4-(1,3-Dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid
1-[4-(1,3-Dimethyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid
Formel:
C₁₆H₁₈F₃N₅O₂ Molecular Weight: 369,342 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05861924 CAS-Nummer: 1006334-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...