3-[5-Phenyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile

Lieferant: Apollo Scientific

PC410390-1G
APOSPC410390-1GEA 516 EUR
APOSPC410390-1G
3-[5-Phenyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
3-[5-Phenyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Formel: C₁₃H₁₀F₃N₃
Molecular Weight: 265.234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558312
CAS-Nummer: 1006334-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR