(1E)-N'-Hydroxy-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanimidamide

Lieferant: Apollo Scientific

PC410391-1G
APOSPC410391-1GEA 516 EUR
APOSPC410391-1G
(1E)-N'-Hydroxy-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanimidamide
(1E)-N'-Hydroxy-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanimidamide
Formel: C₈H₁₁F₃N₄O
Molecular Weight: 236,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20491475
CAS-Nummer: 1006334-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR