{3-[1'-Methyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propyl}amine

Lieferant: Apollo Scientific

PC410392-1G
APOSPC410392-1GEA 516 EUR
APOSPC410392-1G
{3-[1'-Methyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propyl}amine
{3-[1'-Methyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propyl}amine
Formel: C₁₁H₁₄F₃N₅
Molecular Weight: 273,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08556222
CAS-Nummer: 1006336-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR