2-Methyl-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile

Lieferant: Apollo Scientific

PC410393-1G
APOSPC410393-1GEA 386 EUR
APOSPC410393-1G
2-Methyl-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
2-Methyl-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Formel: C₉H₁₀F₃N₃
Molecular Weight: 217,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08556257
CAS-Nummer: 1006336-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR