N-[(1,3-Dimethyl-1H-pyrazol-4-yl)methyl]-3-(trifluoromethyl)aniline

Lieferant: Apollo Scientific

PC410394-1G
APOSPC410394-1GEA 980 EUR
APOSPC410394-1G
N-[(1,3-Dimethyl-1H-pyrazol-4-yl)methyl]-3-(trifluoromethyl)aniline
N-[(1,3-Dimethyl-1H-pyrazol-4-yl)methyl]-3-(trifluoromethyl)aniline
Formel: C₁₃H₁₄F₃N₃
Molecular Weight: 269,266 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04968305
CAS-Nummer: 1006336-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR