3-[1'-Ethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propan-1-amine

Lieferant: Apollo Scientific

PC410395-1G
APOSPC410395-1GEA 516 EUR
APOSPC410395-1G
3-[1'-Ethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propan-1-amine
3-[1'-Ethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propan-1-amine
Formel: C₁₂H₁₆F₃N₅
Lagertemperatur: Raumtemperatur
MDL: MFCD08556271
CAS-Nummer: 1006336-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR