4-Fluoro-N-[(1-methyl-1H-pyrazol-4-yl)methyl]aniline
Lieferant: Apollo Scientific
PC410396-1G
APOSPC410396-1GEA
647
EUR
APOSPC410396-1G
4-Fluoro-N-[(1-methyl-1H-pyrazol-4-yl)methyl]aniline
4-Fluoro-N-[(1-methyl-1H-pyrazol-4-yl)methyl]aniline
Formel:
C₁₁H₁₂FN₃ Molecular Weight: 205,231 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04968280 CAS-Nummer: 1006336-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...