N-[3-Methoxy-4-(2,2,2-trifluoroethoxy)benzyl]-1,3-dimethyl-1H-pyrazol-4-amine

Lieferant: Apollo Scientific

PC410399-1G
APOSPC410399-1GEA 647 EUR
APOSPC410399-1G
N-[3-Methoxy-4-(2,2,2-trifluoroethoxy)benzyl]-1,3-dimethyl-1H-pyrazol-4-amine
N-[3-Methoxy-4-(2,2,2-trifluoroethoxy)benzyl]-1,3-dimethyl-1H-pyrazol-4-amine
Formel: C₁₅H₁₈F₃N₃O₂
Molecular Weight: 329,318 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08556276
CAS-Nummer: 1006340-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR