3-[1'-Ethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propan-1-amine
Lieferant: Apollo Scientific
PC410400-1G
APOSPC410400-1GEA
386
EUR
APOSPC410400-1G
3-[1'-Ethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propan-1-amine
3-[1'-Ethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propan-1-amine
Formel:
C₁₂H₁₆F₃N₅ Molecular Weight: 287,284 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08556281 CAS-Nummer: 1006340-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...