{2-Methyl-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propyl}amine

Lieferant: Apollo Scientific

PC410402-1G
APOSPC410402-1GEA 516 EUR
APOSPC410402-1G
{2-Methyl-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propyl}amine
{2-Methyl-3-[5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propyl}amine
Formel: C₉H₁₄F₃N₃
Molecular Weight: 221,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06805063
CAS-Nummer: 1006342-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR