5-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine

Lieferant: Apollo Scientific

PC410403-1G
APOSPC410403-1GEA 647 EUR
APOSPC410403-1G
5-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine
5-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine
Formel: C₁₃H₁₆F₃N₅
Molecular Weight: 299,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04968858
CAS-Nummer: 1006342-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR