2-[(5-{[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}-4-ethyl-4H-1,2,4-triazol-3-yl)sulphanyl]acetohydrazide
Lieferant: Apollo Scientific
PC410404-1G
APOSPC410404-1GEA
647
EUR
APOSPC410404-1G
2-[(5-{[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}-4-ethyl-4H-1,2,4-triazol-3-yl)sulphanyl]acetohydrazide
2-[(5-{[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]methyl}-4-ethyl-4H-1,2,4-triazol-3-yl)sulphanyl]acetohydrazide
Formel:
C₁₂H₁₅F₄N₇OS Molecular Weight: 381.352 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03423244 CAS-Nummer: 1006343-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...