3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Lieferant: Apollo Scientific
PC410405-1G
APOSPC410405-1GEA
516
EUR
APOSPC410405-1G
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Formel:
C₈H₈F₃N₃ Molecular Weight: 203,164 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05861675 CAS-Nummer: 1006343-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...