1-[(4-Fluorophenoxy)methyl]-3,5-dimethyl-1H-pyrazol-4-amine

Lieferant: Apollo Scientific

PC410406-1G
APOSPC410406-1GEA 516 EUR
APOSPC410406-1G
1-[(4-Fluorophenoxy)methyl]-3,5-dimethyl-1H-pyrazol-4-amine
1-[(4-Fluorophenoxy)methyl]-3,5-dimethyl-1H-pyrazol-4-amine
Formel: C₁₂H₁₄FN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD04967713
CAS-Nummer: 1006344-04-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR