3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanenitrile

Lieferant: Apollo Scientific

PC410407-1G
APOSPC410407-1GEA 516 EUR
APOSPC410407-1G
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanenitrile
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanenitrile
Formel: C₉H₁₀F₃N₃
Molecular Weight: 217,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739386
CAS-Nummer: 1006348-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR